Thermo Scientific™ Precision™ Digital Circulating Water Baths

Precision Digital Circulating Water Baths are microprocessor controlled and combine simplicity with excellent temperature uniformity.

Manufacturer: Thermo Scientific™ 2865

 View more versions of this product

Catalog No. 15-474-101

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Display Digital LED
    Height (English) Exterior 9.5 in.
    Height (Metric) Exterior 24.1cm
    Length (English) Exterior 14 in.
    Length (Metric) Exterior 35.6cm
    Model 260
    Temperature Range (Metric) Ambient +5° to 99.9°C
    Temperature Uniformity ±0.05°C
    Width (English) Exterior 24.5 in.
    Width (English) Interior 15 in.
    Width (Metric) Exterior 62.2cm
    Width (Metric) Interior 38.1cm
    Cabinet Material Expoxy powder-coated stainless steel
    Dimensions (L x W x H) Exterior 14 x 24.5 x 9.5 in. (35.6 x 62.2 x 24.1cm)
    Electrical Requirements 230V 50/60Hz
    Bath Volume 19L (5.1 gal.)
    Amperage 4.3A
    Control Microprocessor
    Height (English) Interior 7.5 in.
    Height (Metric) Interior 19cm
    Hertz 50/60Hz
    Shipping Weight (English) 45 lb.
    Shipping Weight (Metric) 20.4kg
    Temperature Resolution ±0.05°C
    Chamber Material Stainless steel
    Description Model 260; 5.1 gal. (19L); 230V
    Certifications/Compliance ASTM™ E715 Class IIA standards for uniformity. UL and CE Listed
    Dimensions (D x W x H) Interior 12 x 15 x 7.5 in. (30.5 x 38.1 x 19cm)
    Voltage 230V
    Depth (English) Interior 12 in.
    Depth (Metric) Interior 30.5cm

    Thermo Scientific™ Precision Digital Circulating Water Baths are microprocessor controlled and combine simplicity with excellent temperature uniformity. Ideal for applications where temperature uniformity and control are critical, such as enzymes and serology. Available in three different models, these high performance baths include a stainless-steel gable cover and diffuser shelf.

    • Temperature controller provides ±0.05°C uniformity at 37°C and control resolution of ±0.05°C with stainless-steel gable cover
    • Microprocessor control with digital LED display
    • Set temperature in 0.1°C increments
    • Water flow directed around bath perimeter
    • Elevated sample platform for more thorough circulation
    • Overtemperature protection capability
    • Single switch control for heater and circulator pump
    • Stainless-steel construction; exterior finished with baked-on epoxy-polyester

    Model 260

    • Capacity: 5.1 gal. (19L)

    Model 265

    • Capacity: 9.1 gal. (34.5L)

    Model 270

    • Capacity: 23.8 gal. (89L)

    Includes:

    Stainless-steel gable cover; Diffuser shelf; 3-wire cord and plug; Duck

    Recommended for:

    • Bacteriological Examinations Coagulation Tests Coliform Determinations Copper Strip Corrosion Tests Crude Oil Studies Cytochemistry
    • Demulsibility Studies Enzyme Studies
    • Environmental Studies Food Processing QC Genetic Studies Hormone Studies Immunological Research Incubation for Microbiological Assays
    • Metallurgical Analysis Molecular Biology Protein Analysis Radioactive Isotope Uptake Studies Radiochemistry Serological Research Thawing Thawing Blood
    • Tissue Culture Research Virology Research Warming Reagents Water Quality Research

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit