Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ Digital Circulating Water Baths

Precision Digital Circulating Water Baths are microprocessor controlled and combine simplicity with excellent temperature uniformity.

Manufacturer:  Thermo Scientific™ 2866

 View more versions of this product

Catalog No. 15-474-106

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Thermo Scientific™ Precision Digital Circulating Water Baths are microprocessor controlled and combine simplicity with excellent temperature uniformity. Ideal for applications where temperature uniformity and control are critical, such as enzymes and serology. Available in three different models, these high performance baths include a stainless-steel gable cover and diffuser shelf.

  • Temperature controller provides ±0.05°C uniformity at 37°C and control resolution of ±0.05°C with stainless-steel gable cover
  • Microprocessor control with digital LED display
  • Set temperature in 0.1°C increments
  • Water flow directed around bath perimeter
  • Elevated sample platform for more thorough circulation
  • Overtemperature protection capability
  • Single switch control for heater and circulator pump
  • Stainless-steel construction; exterior finished with baked-on epoxy-polyester

Model 260

  • Capacity: 5.1 gal. (19L)

Model 265

  • Capacity: 9.1 gal. (34.5L)

Model 270

  • Capacity: 23.8 gal. (89L)

Includes:

Stainless-steel gable cover; Diffuser shelf; 3-wire cord and plug; Duck

Recommended for:

  • Bacteriological Examinations Coagulation Tests Coliform Determinations Copper Strip Corrosion Tests Crude Oil Studies Cytochemistry
  • Demulsibility Studies Enzyme Studies
  • Environmental Studies Food Processing QC Genetic Studies Hormone Studies Immunological Research Incubation for Microbiological Assays
  • Metallurgical Analysis Molecular Biology Protein Analysis Radioactive Isotope Uptake Studies Radiochemistry Serological Research Thawing Thawing Blood
  • Tissue Culture Research Virology Research Warming Reagents Water Quality Research
Description & Specifications

Specifications

Ambient +5° to 99.9°C
120V
Microprocessor
30.5cm
9.5 in.
24.1cm
50/60Hz
35.6cm
68 lb.
±0.05°C
36.5 in.
92.7cm
Expoxy powder-coated stainless steel
Model 265; Capacity: 34.5L; 120V
14 x 36.5 x 9.5 in. (35.6 x 92.7 x 24.1cm)
120V 50/60Hz
34.5L (9.1 gal.)
12.9A
12 in.
Digital LED
7.5 in.
19cm
14 in.
265
30.8kg
±0.05°C
27 in.
68.6cm
Stainless steel
ASTM? E715 Class IIA standards for uniformity. UL and CE Listed
12 x 27 x 7.5 in. (30.5 x 68.6 x 19cm)
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit