Thermo Scientific™ Precision™ Digital Circulating Water Baths

Precision Digital Circulating Water Baths are microprocessor controlled and combine simplicity with excellent temperature uniformity.

Manufacturer: Thermo Scientific™ 2868

 View more versions of this product

Catalog No. 15-474-105

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Model 270
    Shipping Weight (Metric) 38.5kg
    Temperature Resolution ±0.05°C
    Cabinet Material Expoxy powder-coated stainless steel
    Certifications/Compliance ASTM™ E715 Class IIA standards for uniformity. UL and CE Listed
    Amperage 12.9A
    Display Digital LED
    Height (English) Exterior 12 in.
    Height (English) Interior 9.5 in.
    Height (Metric) Exterior 30cm
    Height (Metric) Interior 24cm
    Length (Metric) Exterior 51cm
    Temperature Range (Metric) Ambient +5° to 99.9°C
    Temperature Uniformity ±0.05°C
    Chamber Material Stainless steel
    Description Model 270; 23.8 gal. (89L); 120V
    Dimensions (D x W x H) Interior 18 x 36 x 9.5 in. (46 x 91 x 24cm)
    Electrical Requirements 120V 50/60Hz
    Voltage 120V
    Depth (English) Interior 18 in.
    Depth (Metric) Interior 46cm
    Hertz 50/60Hz
    Length (English) Exterior 20 in.
    Shipping Weight (English) 85 lb.
    Width (English) Exterior 45 in.
    Width (English) Interior 36 in.
    Width (Metric) Exterior 114cm
    Width (Metric) Interior 91cm
    Dimensions (L x W x H) Exterior 20 x 45 x 12 in. (51 x 114 x 30cm)
    Bath Volume 89L (23.8 gal.)
    Control Microprocessor

    Thermo Scientific™ Precision Digital Circulating Water Baths are microprocessor controlled and combine simplicity with excellent temperature uniformity. Ideal for applications where temperature uniformity and control are critical, such as enzymes and serology. Available in three different models, these high performance baths include a stainless-steel gable cover and diffuser shelf.

    • Temperature controller provides ±0.05°C uniformity at 37°C and control resolution of ±0.05°C with stainless-steel gable cover
    • Microprocessor control with digital LED display
    • Set temperature in 0.1°C increments
    • Water flow directed around bath perimeter
    • Elevated sample platform for more thorough circulation
    • Overtemperature protection capability
    • Single switch control for heater and circulator pump
    • Stainless-steel construction; exterior finished with baked-on epoxy-polyester

    Model 260

    • Capacity: 5.1 gal. (19L)

    Model 265

    • Capacity: 9.1 gal. (34.5L)

    Model 270

    • Capacity: 23.8 gal. (89L)

    Includes:

    Stainless-steel gable cover; Diffuser shelf; 3-wire cord and plug; Duck

    Recommended for:

    • Bacteriological Examinations Coagulation Tests Coliform Determinations Copper Strip Corrosion Tests Crude Oil Studies Cytochemistry
    • Demulsibility Studies Enzyme Studies
    • Environmental Studies Food Processing QC Genetic Studies Hormone Studies Immunological Research Incubation for Microbiological Assays
    • Metallurgical Analysis Molecular Biology Protein Analysis Radioactive Isotope Uptake Studies Radiochemistry Serological Research Thawing Thawing Blood
    • Tissue Culture Research Virology Research Warming Reagents Water Quality Research

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit