Thermo Scientific™ Precision™ Digital Coliform Water Baths

Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination and feature factory-preset temperatures for easy use.

Manufacturer: Thermo Scientific™ 2860

 View more versions of this product

Catalog No. 15-459-12

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Model 251
    Shipping Weight (English) 35 lb.
    Shipping Weight (Metric) 15.8kg
    Temperature Resolution ±0.05°C
    Width (English) Interior 12 in.
    Width (Metric) Exterior 39.4cm
    Dimensions (L x W x H) Exterior 18.5 x 15.5 x 9.5 in. (47 x 39.4 x 24.1cm)
    Voltage 115V
    Depth (Metric) Interior 35.6cm
    Height (English) Exterior 9.5 in.
    Height (English) Interior 8 in.
    Height (Metric) Exterior 24.1cm
    Height (Metric) Interior 20.3cm
    Length (English) Exterior 18.5 in.
    Length (Metric) Exterior 47cm
    Temperature Presets 35, 41.5, 44.5, 45.5°C
    Temperature Range (Metric) 35° to 45.5°C
    Temperature Uniformity ±0.05°C
    Width (English) Exterior 15.5 in.
    Capacity (Metric) 17.5L
    Certifications/Compliance UL listed
    Electrical Requirements 115V 50/60Hz
    Dimensions (D x W x H) Interior 14 x 12 x 8 in. (35.6 x 30.4 x 20.3cm)
    Depth (English) Interior 14 in.
    Hertz 50/60Hz
    Width (Metric) Interior 30.4cm
    Capacity (English) 4.8 gal.
    Description 4.8 gal.; 115V 50/60Hz
    Capacity 4.8 gal., 17.5L
    Cabinet Material Epoxy powder-coated stainless steel
    Chamber Material Stainless steel

    Thermo Scientific™ Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination. Baths feature factory-preset temperatures for easy use, even by untrained operators.

    • Temperature controller provides ±0.05°C uniformity and ±0.05°C control resolution with stainless-steel gable cover
    • Microprocessor control with digital LED display
    • Water pump gently directs water flow around bath perimeter for nonturbulent agitation
    • Overtemperature protection capability
    • Rugged stainless-steel chamber with easy to clean epoxy powder-coated exterior

    Includes:

    Stainless-steel gable cover, diffuser shelf and duck

    Recommended for:

    • Bacteriological Examinations
    • Coliform Determinations
    • Environmental Studies
    • Food Processing QC
    • Incubation for Microbiological Assays
    • Water Quality Research

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit