Thermo Scientific™ Precision™ Digital Coliform Water Baths

Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination and feature factory-preset temperatures for easy use.

Manufacturer: Thermo Scientific™ 2861

 View more versions of this product

Catalog No. 15-459-13

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Height (English) Interior 8 in.
    Height (Metric) Interior 20.3cm
    Model 251
    Shipping Weight (English) 35 lb.
    Temperature Range (Metric) 35° to 45.5°C
    Temperature Resolution ±0.05°C
    Temperature Uniformity ±0.05°C
    Width (English) Interior 12 in.
    Capacity (Metric) 17.5L
    Voltage 230V
    Chamber Material Stainless steel
    Depth (English) Interior 14 in.
    Height (Metric) Exterior 24.1cm
    Hertz 50/60Hz
    Length (English) Exterior 18.5 in.
    Length (Metric) Exterior 47cm
    Shipping Weight (Metric) 15.8kg
    Temperature Presets 35, 41.5, 44.5, 45.5°C
    Width (English) Exterior 15.5 in.
    Width (Metric) Exterior 39.4cm
    Width (Metric) Interior 30.4cm
    Capacity (English) 4.8 gal.
    Certifications/Compliance UL listed
    Capacity 4.8 gal., 17.5L
    Dimensions (L x W x H) Exterior 18.5 x 15.5 x 9.5 in. (47 x 39.4 x 24.1cm)
    Electrical Requirements 230V 50/60Hz
    Dimensions (D x W x H) Interior 14 x 12 x 8 in. (35.6 x 30.4 x 20.3cm)
    Description 4.8 gal.; 230V 50/60Hz
    Cabinet Material Epoxy powder-coated stainless steel
    Depth (Metric) Interior 35.6cm
    Height (English) Exterior 9.5 in.

    Thermo Scientific™ Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination. Baths feature factory-preset temperatures for easy use, even by untrained operators.

    • Temperature controller provides ±0.05°C uniformity and ±0.05°C control resolution with stainless-steel gable cover
    • Microprocessor control with digital LED display
    • Water pump gently directs water flow around bath perimeter for nonturbulent agitation
    • Overtemperature protection capability
    • Rugged stainless-steel chamber with easy to clean epoxy powder-coated exterior

    Includes:

    Stainless-steel gable cover, diffuser shelf and duck

    Recommended for:

    • Bacteriological Examinations
    • Coliform Determinations
    • Environmental Studies
    • Food Processing QC
    • Incubation for Microbiological Assays
    • Water Quality Research

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit