Thermo Scientific™ Precision™ Digital Coliform Water Baths

Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination and feature factory-preset temperatures for easy use.

Manufacturer: Thermo Scientific™ 2862

 View more versions of this product

Catalog No. 15-459-15

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Hertz 50/60Hz
    Model 253
    Shipping Weight (Metric) 27.2kg
    Temperature Presets 35, 41.5, 44.5, 45.5°C
    Width (Metric) Exterior 91.4cm
    Capacity (Metric) 34.5L
    Description 9.1 gal.; 115V 50/60Hz
    Certifications/Compliance UL listed
    Dimensions (L x W x H) Exterior 14 x 36 x 9.5 in. (35.6 x 91.4 x 24.1cm)
    Voltage 115V
    Cabinet Material Epoxy powder-coated stainless steel
    Depth (English) Interior 12 in.
    Height (English) Interior 7.5 in.
    Height (Metric) Exterior 24.1cm
    Height (Metric) Interior 19cm
    Shipping Weight (English) 60 lb.
    Temperature Range (Metric) 35° to 45.5°C
    Temperature Resolution ±0.05°C
    Temperature Uniformity ±0.05°C
    Width (English) Exterior 36 in.
    Width (English) Interior 27 in.
    Width (Metric) Interior 68.6cm
    Capacity (English) 9.1 gal.
    Capacity 9.1 gal., 34.5L
    Electrical Requirements 115V 50/60Hz
    Dimensions (D x W x H) Interior 12 x 27 x 7.5 in. (30.5 x 68.6 x 19cm)
    Chamber Material Stainless steel
    Height (English) Exterior 9.5 in.
    Length (English) Exterior 14 in.
    Length (Metric) Exterior 35.6cm
    Depth (Metric) Interior 30.5cm

    Thermo Scientific™ Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination. Baths feature factory-preset temperatures for easy use, even by untrained operators.

    • Temperature controller provides ±0.05°C uniformity and ±0.05°C control resolution with stainless-steel gable cover
    • Microprocessor control with digital LED display
    • Water pump gently directs water flow around bath perimeter for nonturbulent agitation
    • Overtemperature protection capability
    • Rugged stainless-steel chamber with easy to clean epoxy powder-coated exterior

    Includes:

    Stainless-steel gable cover, diffuser shelf and duck

    Recommended for:

    • Bacteriological Examinations
    • Coliform Determinations
    • Environmental Studies
    • Food Processing QC
    • Incubation for Microbiological Assays
    • Water Quality Research

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit