Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ Digital Coliform Water Baths

Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination and feature factory-preset temperatures for easy use.

Specifications

Material Epoxy Powder-coated Steel
Height (English) 9.5 in.
Height (Metric) 24.1 cm
Frequency 50/60 Hz
Temperature Presets 35, 41.5, 44.5, 45.5°C
View More Specs

Products 1
Catalog Number Mfr. No. Voltage Frequency Price Quantity & Availability  
Catalog Number Mfr. No. Voltage Frequency Price Quantity & Availability  
15-459-15
Accessories View Documents
Thermo Scientific™
2862
115 V 50/60 Hz This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Thermo Scientific™ Precision Digital Coliform Water Baths are designed specifically for fecal coliform determination. Baths feature factory-preset temperatures for easy use, even by untrained operators.

  • Temperature controller provides ±0.05°C uniformity and ±0.05°C control resolution with stainless-steel gable cover
  • Microprocessor control with digital LED display
  • Water pump gently directs water flow around bath perimeter for nonturbulent agitation
  • Overtemperature protection capability
  • Rugged stainless-steel chamber with easy to clean epoxy powder-coated exterior

Includes:

Stainless-steel gable cover, diffuser shelf and duck

Recommended for:

  • Bacteriological Examinations
  • Coliform Determinations
  • Environmental Studies
  • Food Processing QC
  • Incubation for Microbiological Assays
  • Water Quality Research
Specifications

Specifications

Epoxy Powder-coated Steel
24.1 cm
35, 41.5, 44.5, 45.5°C
±0.05°C
9.5 in.
50/60 Hz
±0.05°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit