Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ Dubnoff™ Reciprocal Shaking Baths

Precision Dubnoff Reciprocal Shaking Water Baths are designed specifically for applications that require samples to be incubated in a controlled atmosphere.

Manufacturer:  Thermo Scientific™ 2876

 View more versions of this product

Catalog No. 15-453-200

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Description

Thermo Scientific™ Precision Dubnoff Reciprocal Shaking Water Baths are designed specifically for applications that require samples to be incubated in a controlled atmosphere. Provides excellent performance and reliability for a variety of laboratory applications.

  • Equipped with one large and two small gassing hoods
  • Microprocessor temperature and shaker control with digital LED display
  • Adjustable shaking speed from 30 to 200 (OPM) oscillations/min.
  • Push-button convenience for entry of temperature and shaking speed
  • Overtemperature protection capability
  • Automatic motor shutdown capability
  • One-piece, easy-to-remove tray for easy use and maintenance

Ordering Information:

Optional O2/N2/CO2 flowmeter available

Includes:

  • One large and two small gassing hoods
  • Stainless-steel gable cover
  • Duck

Recommended for:

  • Coagulation tests
  • Copper strip erosion tests
  • Cytochemistry
  • Dialysis
  • Enzyme studies
  • Environmental studies
  • Genetic studies
  • Hormone studies
  • Immunological research
  • Incubation for microcentrifugation tubes
  • Metallurgical analysis
  • Molecular biology
  • Radioactive isotope uptake studies
  • Serological research
  • Thawing
  • Thawing blood
  • Tissue culture research
  • Virology research
  • Water quality research
Description & Specifications

Specifications

Ambient to 99.9°C
9.5 in.
24.1cm
50/60Hz
11.5 in.
35.6cm
1.3 to 3.8cm
24.9kg
1000w
±0.05°C
1000w
15 in.
62.2cm
27.9cm
3.9 gal.
Microprocessor
19cm
ASTM? Method E 715 Class IIA standards. UL listed
120V 50/60Hz
120V
6.5 in.
16.5cm
14 in.
0.5 to 1.5 in.
29.2cm
55 lb.
3415 BTU
Reciprocating
±0.05 at 37°C
24.5 in.
11 in.
38.1cm
8.2A
14.5L
7.5 in.
Precision Dubnoff Reciprocal Shaking Baths
14 x 24.5 x 9.5 in. (35.6 x 62.2 x 24.1cm)
3.9 gal., 14.5L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit