Thermo Scientific™ Precision™ General-Purpose Water Baths

Precision General-Purpose Water Baths consist of eight models, including dual chamber and shallow form, with analog or digital control.

Manufacturer: Thermo Scientific™ 2825

 View more versions of this product

Catalog No. 15-474-65

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Height (Metric) Exterior 17.1cm
    Hertz 50/60Hz
    Length (English) Exterior 8.9 in.
    Length (Metric) Exterior 22.3cm
    Model 280 Shallow Chamber
    Shipping Weight (English) 12 lb.
    Shipping Weight (Metric) 5.4kg
    Temperature Range (Metric) Ambient to 95° or 99°C
    Temperature Resolution ±0.1†°C
    Temperature Uniformity ±0.2°C
    Wattage 225w
    Width (English) Exterior 14 in.
    Width (English) Interior 11.5 in.
    Width (Metric) Exterior 35.6cm
    Width (Metric) Interior 29.2cm
    Amperage 2A
    Capacity (Metric) 1.5L
    Control Digital
    Length (English) Interior 6 in.
    Length (Metric) Interior 15.2cm
    Operating Temperature 95°C
    Certifications/Compliance UL listed
    Dimensions (L x W x H) Exterior 8.9 x 14 x 6.7 in. (22.3 x 35.6 x 17.1cm)
    Electrical Requirements 120V 50/60Hz
    Voltage 120V
    Cabinet Material Epoxy powder-coated stainless steel
    Chamber Material Stainless steel
    Depth (English) Interior 2 in.
    Depth (Metric) Interior 5.1cm
    Height (English) Exterior 6.7 in.

    Thermo Scientific™ Precision General-Purpose Water Baths consist of eight models, including dual chamber and shallow form, with analog or digital control. Easy-to-maintain, seamless-stainless steel interior chamber and epoxy powder-coated exterior are resistant to corrosion and chemical damage.

    Rugged, high-performance baths maintain water temperature from ambient to 95°C or 99.9°C (depending on model) with ±0.2°C uniformity (at 37°C) and ±0.1°C control resolution (at 37°C) with stainless-steel gable cover.

    • Choice of analog or digital temperature control
    • Chamber capacities ranging from 1.5 to 43L
    • Front-mounted controls simplify operation
    • Overtemperature safety circuitry prevents thermal runaway
    • Dual-chamber models have independent controls for operating at different temperatures

    Analog Baths

    • Easy-to-use analog controls and hydraulic thermostat are ideal for applications in which temperature setpoints seldom change
    • Backup high-limit safety thermostat
    • Illuminated power switch

    Digital Baths

    • Allow temperature setpoint with ±0.1°C precision at the push of a button
    • Microprocessor control with bright three-digit LED temperature readout
    • Automatic overtemperature protection

    Includes:

    Gable cover: stainless steel for all except Models 181 and 281, which are polypropylene; Diffuser shelf; Spirit-filled thermometer (analog baths only); Rubber duck

    Recommended for:

    • Bacteriological examinations
    • Coagulation tests
    • Copper strip corrosion tests
    • Crude oil studies
    • Cytochemistry
    • Demulsibility studies
    • Environmental studies
    • Food processing QC
    • Genetic studies
    • Hormone studies
    • Immunological research
    • Incubation for microbiological assays
    • Incubation for microcentrifugation tubes
    • Melting agar
    • Metallurgical analysis
    • Molecular biology
    • Protein analysis
    • Radioactive isotope uptake studies
    • Radiochemistry
    • Serological research
    • Thawing
    • Thawing blood
    • Thawing cryopreservation vials
    • Tissue culture research
    • Virology research
    • Warming reagents
    • Water quality research

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit