Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Precision™ General-Purpose Water Baths

Precision General-Purpose Water Baths consist of eight models, including dual chamber and shallow form, with analog or digital control.

Manufacturer:  Thermo Scientific™ 2832

 View more versions of this product

Catalog No. 15-474-16

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Description

Thermo Scientific™ Precision General-Purpose Water Baths consist of eight models, including dual chamber and shallow form, with analog or digital control. Easy-to-maintain, seamless-stainless steel interior chamber and epoxy powder-coated exterior are resistant to corrosion and chemical damage.

Rugged, high-performance baths maintain water temperature from ambient to 95°C or 99.9°C (depending on model) with ±0.2°C uniformity (at 37°C) and ±0.1°C control resolution (at 37°C) with stainless-steel gable cover.
  • Choice of analog or digital temperature control
  • Chamber capacities ranging from 1.5 to 43L
  • Front-mounted controls simplify operation
  • Overtemperature safety circuitry prevents thermal runaway
  • Dual-chamber models have independent controls for operating at different temperatures

Analog Baths

  • Easy-to-use analog controls and hydraulic thermostat are ideal for applications in which temperature setpoints seldom change
  • Backup high-limit safety thermostat
  • Illuminated power switch

Digital Baths

  • Allow temperature setpoint with ±0.1°C precision at the push of a button
  • Microprocessor control with bright three-digit LED temperature readout
  • Automatic overtemperature protection

Includes:

Gable cover: stainless steel for all except Models 181 and 281, which are polypropylene; Diffuser shelf; Spirit-filled thermometer (analog baths only); Rubber duck

Recommended for:

  • Bacteriological examinations
  • Coagulation tests
  • Copper strip corrosion tests
  • Crude oil studies
  • Cytochemistry
  • Demulsibility studies
  • Environmental studies
  • Food processing QC
  • Genetic studies
  • Hormone studies
  • Immunological research
  • Incubation for microbiological assays
  • Incubation for microcentrifugation tubes
  • Melting agar
  • Metallurgical analysis
  • Molecular biology
  • Protein analysis
  • Radioactive isotope uptake studies
  • Radiochemistry
  • Serological research
  • Thawing
  • Thawing blood
  • Thawing cryopreservation vials
  • Tissue culture research
  • Virology research
  • Warming reagents
  • Water quality research
Description & Specifications

Specifications

Ambient to 95° or 99°C
Epoxy powder-coated steel
6 in.
9.75 in.
50/60Hz
41cm
16 lb.
±0.1†°C
300w
6 in.
15.2cm
5.5L
11.5 in.
99.9°C
16.2 x 8 x 9.75 in. (41 x 20.3 x 24.8cm)
230V
Stainless steel
15.2cm
24.8cm
16.2 in.
182
7.3kg
±0.2°C
8 in.
20.3cm
1.3A
Analog
29.2cm
UL listed
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit