Learn More
Thermo Scientific™ Precision™ Shaking Water Bath Bundle PROMO
Shaking water bath supports a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing. Bundle includes one shaking water bath along with one general purpose water bath.
$3350.00
Specifications
Chamber Material 304 Stainless Steel
Controller Type Digital
Display Monochrome LCD
Temperature Presets 4
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
TSSWB15PMO
View Documents Promotion Details
Thermo Scientific™
TSSWB15PMO
15 L Precision™ Shaking Water Bath, 5 L General Purpose Water Bath
Each for $3,350.00
Add to cart
TSSWB27PMO
View Documents Promotion Details
Thermo Scientific™
TSSWB27PMO
27 L Precision™ Shaking Water Bath, 5 L General Purpose Water Bath
Each for $3,350.00
Add to cart
Description
Description
  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
Specifications
Specifications
304 Stainless Steel
Digital
Monochrome LCD
4