Thermo Scientific™ Precision™ Shallow-Form Reciprocal Shaking Baths

Precision Shallow-Form Reciprocal Shaking Water Baths offer push-button convenience for entry of temperature and shaking speed. Water baths provide excellent performance and reliability.

Manufacturer: Thermo Scientific™ 2874

 View more versions of this product

Catalog No. 15-453-205

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Height (Metric) Removable Tray 8.9cm
    Electrical Requirements 120V 50/60Hz
    Capacity 3.9 gal., 14.5L
    Hertz 50/60Hz
    Length (English) Exterior 14 in.
    Length (English) Removable Tray 11.5 in.
    Length (English) Stroke 0.5 to 1.5 in.
    Length (Metric) Exterior 35.6cm
    Length (Metric) Removable Tray 29.2cm
    Length (Metric) Stroke 1.3 to 3.8cm
    Net Weight (English) 55 lb.
    Temperature Range (Metric) Ambient to 99.9°C
    Width (English) Removable Tray 11 in.
    Width (Metric) Exterior 62.2cm
    Width (Metric) Interior 38.1cm
    Capacity (English) 3.9 gal.
    Capacity (Metric) 14.5L
    Depth (English) Interior 7.5 in.
    Depth (Metric) Interior 19cm
    Description Capacity: 14.5L; 120V; 50/60Hz; 8.3A
    Length (English) Interior 12 in.
    Length (Metric) Interior 30.5cm
    Voltage 120V
    Height (English) Exterior 9.5 in.
    Height (English) Removable Tray 3.5 in.
    Net Weight (Metric) 24.9kg
    Shaking Motion Reciprocating
    Temperature Resolution ±0.05°C
    Temperature Uniformity ±0.05 at 37°C
    Width (English) Exterior 24.5 in.
    Width (English) Interior 15 in.
    Width (Metric) Removable Tray 27.9cm
    Amperage 8.3A
    Control Microprocessor
    Oscillations per Minute 30 to 200
    Certifications/Compliance ASTM™ Method E 715 Class IIA standards. UL listed
    Dimensions (L x W x H) Exterior 14 x 24.5 x 9.5 in. (35.6 x 62.2 x 24.1cm)
    Height (Metric) Exterior 24.1cm

    Thermo Scientific™ Precision Shallow-Form Reciprocal Shaking Water Baths offer push-button convenience for entry of temperature and shaking speed. Water baths provide excellent performance and reliability for a variety of laboratory applications.

    • Microprocessor temperature and shaker control with digital LED display
    • Adjustable shaking speed from 30 to 200 oscillations per minute
    • Push-button convenience for entry of temperature and shaking speed
    • Overtemperature protection capability
    • Automatic motor shutdown capability
    • One-piece, easy-to-remove tray for easy use and maintenance
    • Corrosion-resistant, stainless-steel interior and exterior

    Ordering Information:

    Optional accessories include flowmeter and gassing hoods.

    Includes:

    • Stainless-steel gable cover
    • 230V model includes European-style 50Hz power cord; standard 220V 60Hz power cord (3176836) available separately
    • Duck

    Recommended for:

    • Coagulation tests
    • Copper strip erosion tests
    • Cytochemistry
    • Dialysis
    • Enzyme studies
    • Environmental studies
    • Genetic studies
    • Hormone studies
    • Immunological research
    • Incubation for microcentrifugation tubes
    • Metallurgical analysis
    • Molecular biology
    • Radioactive isotope uptake studies
    • Serological research
    • Thawing
    • Thawing blood
    • Tissue culture research
    • Virology research
    • Water quality research

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit