Thermo Scientific™ Precision™ Shallow-Form Reciprocal Shaking Baths

Precision Shallow-Form Reciprocal Shaking Water Baths offer push-button convenience for entry of temperature and shaking speed. Water baths provide excellent performance and reliability.

Manufacturer: Thermo Scientific™ 2875

 View more versions of this product

Catalog No. 15-453-201B

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Length (English) Removable Tray 11.5 in.
    Net Weight (English) 55 lb.
    Width (Metric) Interior 38.1cm
    Length (English) Interior 12 in.
    Dimensions (L x W x H) Exterior 14 x 24.5 x 9.5 in. (35.6 x 62.2 x 24.1cm)
    Capacity 3.9 gal., 14.5L
    Hertz 50/60Hz
    Length (Metric) Stroke 1.3 to 3.8cm
    Temperature Range (Metric) Ambient to 99.9°C
    Net Weight (Metric) 24.9kg
    Shaking Motion Reciprocating
    Width (English) Interior 15 in.
    Width (English) Removable Tray 11 in.
    Width (Metric) Exterior 62.2cm
    Amperage 4.3A
    Capacity (English) 3.9 gal.
    Capacity (Metric) 14.5L
    Control Microprocessor
    Depth (English) Interior 7.5 in.
    Depth (Metric) Interior 19cm
    Description Capacity: 14.5L; 230V; 50/60Hz; 4.3A
    Oscillations per Minute 30 to 200
    Voltage 230V
    Certifications/Compliance ASTM™ Method E 715 Class IIA standards. UL listed
    Height (English) Exterior 9.5 in.
    Height (Metric) Exterior 24.1cm
    Height (Metric) Removable Tray 8.9cm
    Length (English) Exterior 14 in.
    Length (English) Stroke 0.5 to 1.5 in.
    Length (Metric) Exterior 35.6cm
    Length (Metric) Removable Tray 29.2cm
    Temperature Resolution ±0.05°C
    Temperature Uniformity ±0.05 at 37°C
    Width (English) Exterior 24.5 in.
    Width (Metric) Removable Tray 27.9cm
    Length (Metric) Interior 30.5cm
    Electrical Requirements 230V 50/60Hz
    Height (English) Removable Tray 3.5 in.

    Thermo Scientific™ Precision Shallow-Form Reciprocal Shaking Water Baths offer push-button convenience for entry of temperature and shaking speed. Water baths provide excellent performance and reliability for a variety of laboratory applications.

    • Microprocessor temperature and shaker control with digital LED display
    • Adjustable shaking speed from 30 to 200 oscillations per minute
    • Push-button convenience for entry of temperature and shaking speed
    • Overtemperature protection capability
    • Automatic motor shutdown capability
    • One-piece, easy-to-remove tray for easy use and maintenance
    • Corrosion-resistant, stainless-steel interior and exterior

    Ordering Information:

    Optional accessories include flowmeter and gassing hoods.

    Includes:

    • Stainless-steel gable cover
    • 230V model includes European-style 50Hz power cord; standard 220V 60Hz power cord (3176836) available separately
    • Duck

    Recommended for:

    • Coagulation tests
    • Copper strip erosion tests
    • Cytochemistry
    • Dialysis
    • Enzyme studies
    • Environmental studies
    • Genetic studies
    • Hormone studies
    • Immunological research
    • Incubation for microcentrifugation tubes
    • Metallurgical analysis
    • Molecular biology
    • Radioactive isotope uptake studies
    • Serological research
    • Thawing
    • Thawing blood
    • Tissue culture research
    • Virology research
    • Water quality research

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit