Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prednisolone Sodium Phosphate, USP, 96-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
125-02-0
C21H27Na2O8P
484.39
VJZLQIPZNBPASX-UHFFFAOYNA-L
disodium 1,10-dihydroxy-9a,11a-dimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
[Na+].[Na+].CC12CC(O)C3C(CCC4=CC(=O)C=CC34C)C1CCC2(O)C(=O)COP([O-])([O-])=O
125-02-0
C21H27Na2O8P
484.39
VJZLQIPZNBPASX-UHFFFAOYNA-L
disodium 1,10-dihydroxy-9a,11a-dimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
[Na+].[Na+].CC12CC(O)C3C(CCC4=CC(=O)C=CC34C)C1CCC2(O)C(=O)COP([O-])([O-])=O

P1417, 125-02-0, C21H27Na2O8P

Spectrum™ Prednisolone Sodium Phosphate, USP is used to treat conditions such as arthritis, blood problems, immune system disorders, skin and eye conditions, breathing problems, cancer, and severe allergies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Specific Rotation +95° to +102°
Water 0.065
pH 7.5 to 10.5
Quantity 100 g
CAS Max % 1
Percent Purity 96 to 102%
Grade USP