Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prednisone, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
53-03-2
C21H26O5
358.434
XOFYZVNMUHMLCC-ZPOLXVRWSA-N
prednisone,deltasone,dehydrocortisone,decortin,meticorten,orasone,prednison,rectodelt,ultracorten,dacortin
5865
CHEBI:8382
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
CC12CC(=O)C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C
53-03-2
C21H26O5
358.434
XOFYZVNMUHMLCC-ZPOLXVRWSA-N
prednisone,deltasone,dehydrocortisone,decortin,meticorten,orasone,prednison,rectodelt,ultracorten,dacortin
5865
CHEBI:8382
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
CC12CC(=O)C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C
Packaging Glass Bottle
Quantity 25g
Melting Point 225.0°C to 229.0°C
Assay Percent Range 98%
Formula Weight 358.43
Chemical Name or Material Prednisone