Learn More
Contec™ PREempt™ RTU Disinfectant Solution
Cleans and disinfects environmental surfaces in cleanrooms and laboratory areas including work stations, fume hoods, equipment and other hard non-porous environmental surfaces. Contec™ PREempt™ RTU Disinfectant Solution helps to ensure user, protocol and product compliance with fast contact times and broad-spectrum efficacy.
Supplier: Contec™ 21105
This item is not returnable.
View return policy
Description
- AHP - Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase potency and cleaning performance
- Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
- Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
- Responsible - Designed to be easier on surfaces, staff and occupants. Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
- 5 minute tuberculocidal
- 10 minute fungicidal
- 30 seconds broad-spectrum sanitizing
- 1 minute bactericidal, virucidal
Specifications
Disinfectant Solution | |
Bottle | |
1 gal. | |
3.8 L |