Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pref-1/DLK1/FA1, Rat anti-Mouse, Clone: PF105B, Novus Biologicals™

Rat Monoclonal Antibody

$240.50 - $366.50

Specifications

Antigen Pref-1/DLK1/FA1
Clone PF105B
Applications Western Blot, ELISA
Host Species Rat
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013164
View Documents
Novus Biologicals
NBP280044100UG
100 ug
Each for $366.50
Add to cart
 
NB013165
View Documents
Novus Biologicals
NBP28004450UG
50 ug
Each for $240.50
Add to cart
 
Description

Description

Pref-1/DLK1/FA1 Monoclonal antibody specifically detects Pref-1/DLK1/FA1 in Mouse Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Pref-1/DLK1/FA1
Western Blot, ELISA
Mouse
Primary
PF105B
Rat
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit