Learn More
Labconco Prefilter for Nexus Horizontal Clean Bench
GSA_VASDP
Disposable prefilters trap large particles to extend the HEPA filter life.
$33.00 - $39.15
Specifications
For Use With (Equipment) Enclosures
Product Type Prefilter
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions Price Quantity & Availability  
17-200-221
GSA_VASDP
View Documents
Labconco
5326800
16 x 18 in.
Each for $33.00
Only null left
Add to Cart
17-200-230
GSA_VASDP
View Documents
Labconco
5326801
16 x 24 in.
Each for $38.30
Only null left
Add to Cart
17-200-207
GSA_VASDP
View Documents
Labconco
5326802
16 x 25 in.
Each for $39.15
Only null left
Add to Cart
Description
Description
  • MERV 8 rating
  • Installation required
  • Labconco recommends changing every 3 months, depending on room cleanliness
  • Refer to Nexus user guide to determine quantity and amount for your clean bench
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Enclosures
Prefilter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.