Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRELID1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551228

Catalog No. PA551228


Add to cart

Description

Description

The antibody detects endogenous levels of total PRELID1 protein.

May be important for the development of vital and immunocompetent organs.
Specifications

Specifications

PRELID1
Polyclonal
Unconjugated
PRELID1
Liquid
PRELID1
Fusion protein of human PRELID1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9Y255
PRELI
Rabbit
IgG
100 μL
-20°C
Primary
27166
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit