Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRELP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578631
Description
Positive Control: mouse heart Predicted Reactivity: Human (100%), Rat (91%), Rhesus Monkey (99%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PRELP is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage.
Specifications
PRELP | |
Polyclonal | |
Unconjugated | |
PRELP | |
55 kDa leucine-rich repeat protein of articular cartilage; 7330409J17Rik; MST161; MSTP161; PRELP; Prolargin; prolargin proteoglycan; proline and arginine rich end leucine rich repeat protein; proline arginine-rich end leucine-rich repeat; proline/arginine-rich end leucine-rich repeat protein; proline-arginine-rich end leucine-rich repeat protein; SLRR2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116847, 5549 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.95 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P51888, Q9JK53 | |
PRELP | |
Recombinant protein encompassing a sequence within the center region of human PRELP. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction