Learn More
Nor-Lake™ Scientific Premier Glass Door Pharmacy Refrigerator, Bottom Mount
Feature an all-new temperature display and alarm module offering an array of robust security features with a monitoring and alarm system that is 100% independent of a microprocessor temperature controller for additional product security.
$5761.00 - $13800.00
Specifications
Amperage 6 A
Display Digital
Ports 1
Phase 1 ph
Plug Type NEMA 5-15 P
View More Specs
Products 10
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
22-651-198
View Documents
Nor-Lake™ Scientific
PHPR121WWG/0
12 cu. ft. Glass Solid (Refrigerator)
Each for $5,761.00
Add to cart
22-651-199
View Documents
Nor-Lake™ Scientific
PHPR161WWG/0
16 cu. ft. Glass Swing
Each for $6,791.00
Add to cart
22-651-200
View Documents
Nor-Lake™ Scientific
PHPR231WWG/0
23 cu. ft. Glass Swing
Each for $7,345.00
Add to cart
22-651-201
View Documents
Nor-Lake™ Scientific
PHPR261WWG/0
26 cu. ft. Glass Swing
Each for $7,547.00
Add to cart
22-651-202
View Documents
Nor-Lake™ Scientific
PHPR332WWG/0
33 cu. ft. Glass Sliding
Each for $9,325.00
Add to cart
22-651-203
View Documents
Nor-Lake™ Scientific
PHPR362WWG/0
36 cu. ft. Glass Swing
Each for $9,410.00
Add to cart
22-651-204
View Documents
Nor-Lake™ Scientific
PHPR472WWG/0
47 cu. ft. Glass Sliding
Each for $10,736.00
Add to cart
22-651-205
View Documents
Nor-Lake™ Scientific
PHPR492WWG/0
49 cu. ft. Glass Swing
Each for $10,807.00
Add to cart
22-651-206
View Documents
Nor-Lake™ Scientific
PHPR693WWG/0
69 cu. ft. Glass Sliding
Each for $13,418.00
Add to cart
22-651-207
View Documents
Nor-Lake™ Scientific
PHPR723WWG/0
72 cu. ft. Glass Swing
Each for $13,800.00
Add to cart
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Standard temperature display
  • 2 probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts, keyed door locks, casters, and pharmacy refrigerator/freezer toolkit, temperature logs and CDC approved power cord labels, plus freestanding thermometer with 3 year certificate of calibration
Specifications
Specifications
6 A
1
NEMA 5-15 P
60 Hz
R134a
115 V
Digital
1 ph
High density urethane foam
Audible and visual high and low temperature alarms
Microprocessor