Learn More
Nor-Lake™ Scientific Premier Pharmacy Dual Temperature Combination Series Refrigerator/Freezer
Nor-Lake™ Scientific Premier Pharmacy Dual Temperature Combination Series Refrigerator/Freezer provides solution where space is at premium. They offer 100% independent digital controls, alarms, and chambers.
$3637.00 - $5471.00
Specifications
Display Digital
Frequency 60 Hz
Temperature Range (Refrigerator) 36°F to 46°F
Phase 1 ph
Plug Type NEMA 5-15 P
View More Specs
Products 11
Catalog Number Mfr. No. Capacity (Metric) Freezer Capacity (Metric) Refrigerator Door Style Monitoring Options Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Freezer Capacity (Metric) Refrigerator Door Style Monitoring Options Price Quantity & Availability  
22-651-185
View Documents
Nor-Lake™ Scientific
PHPRF122WWW/0M
48.1 L 297.3 L Solid Swing Audible and visual high and low temperature alarms
Each for $5,329.00
Add to cart
22-651-186
View Documents
Nor-Lake™ Scientific
PHPRF092WWW/0M
118.9 L 130.3 L Solid Swing Audible and visual high and low temperature alarms
Each for $3,765.00
Add to cart
22-651-187
View Documents
Nor-Lake™ Scientific
PHPRF092WWWLH/0M
118.9 L 130.3 L Solid Swing, Left-hinged Audible and visual high and low temperature alarms
Each for $3,988.00
Add to cart
22-651-188
View Documents
Nor-Lake™ Scientific
PHPRF092SSS/0M
118.9 L 127.4 L Stainless Steel Swing Audible and visual high and low temperature alarms
Each for $4,509.00
Add to cart
22-651-189
View Documents
Nor-Lake™ Scientific
PHPRF092SSSLH/0M
118.9 L 127.4 L Stainless Steel Swing (Left hinged) Audible and visual high and low temperature alarms
Each for $4,767.00
Add to cart
22-651-192
View Documents
Nor-Lake™ Scientific
PHPRF122WWG/0M
48.1 L 297.3 L Glass Solid (Refrigerator) Remote alarm contacts (Refrigerator and Freezer)
Each for $5,329.00
Add to cart
22-651-193
View Documents
Nor-Lake™ Scientific
PHPRF092WWG/0M
118.9 L 130.3 L Glass (Refrigerator) Remote alarm contacts (Refrigerator and Freezer)
Each for $3,765.00
Add to cart
22-651-194
View Documents
Nor-Lake™ Scientific
PHPRF092WWGLH/0M
118.9 L 130.3 L Glass, Left Hinged (Refrigerator) Remote alarm contacts (Refrigerator and Freezer)
Each for $3,988.00
Add to cart
22-651-195
View Documents
Nor-Lake™ Scientific
PHPRF072WWG/0M
48.1 L 147.2 L Glass Swing (Refrigerator) Remote alarm contacts (Refrigerator and Freezer)
Each for $3,637.00
Add to cart
22-651-196
View Documents
Nor-Lake™ Scientific
PHPRF122WWG/0
48.1 L 297.3 L Glass Solid (Refrigerator) Audible and visual high and low temperature alarms
Each for $5,471.00
Add to cart
22-651-197
View Documents
Nor-Lake™ Scientific
PHPRF072WWG/0
36.8 L 147.2 L Glass Solid (Refrigerator) Audible and visual high and low temperature alarms
Each for $3,769.00
Add to cart
Description
Description
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Keyed door locks
Specifications
Specifications
Digital
36°F to 46°F
NEMA 5-15 P
Microprocessor
60 Hz
1 ph
R600a