Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Nor-Lake™ Scientific Premier Pharmacy Freestanding Undercounter Freezer
Ideal choice for pharmacies with undercounter and counter top applications with adequate space for ventilation.
Manufacturer:  Nor-Lake™ Scientific PHPF021WWW/0M
 View more versions of this product
Catalog No. 22-651-179
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • 2 probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts
  • Freestanding thermometer with 3 year certificate of calibration
  • Keyed door locks, leveling legs, and pharmacy refrigerator/freezer toolkit
Specifications
Specifications
Solid Swing
Audible and visual high and low temperature alarms
5°F to -13°F
60 Hz
NEMA 5-15 P
106 lb.
R600a
1.7 cu. ft.
16 in.
13.125 in.
1
17.625 in.
23.75 in.
Microprocessor
High Density Urethane Foam
2 Adjustable
115 V
1 A
80 lb.
Digital
Enameled Steel
16 in.
20.375 in.
1 ph
23.75 in.
Manual
24 in.