Learn More
Nor-Lake™ Scientific Premier Pharmacy Freestanding Undercounter Freezer
Ideal choice for pharmacies with undercounter and counter top applications with adequate space for ventilation.
$1590.31 - $1748.21
Specifications
Exterior Finish Enameled Steel
Display Digital
Ports 1
Phase 1 ph
Plug Type NEMA 5-15 P
View More Specs
Products 4
Catalog Number Mfr. No. Defrost Capacity (English) Freezer Door Style Price Quantity & Availability  
Catalog Number Mfr. No. Defrost Capacity (English) Freezer Door Style Price Quantity & Availability  
22-651-179 View Documents Nor-Lake™ Scientific
PHPF021WWW/0M
Manual 1.7 cu. ft. Solid Swing Each for $1,590.31
Add to cart
22-651-180 View Documents Nor-Lake™ Scientific
PHPF021WWWLH/0M
Manual 1.7 cu. ft. Solid swing door-Left Hinged Each for $1,608.89
Add to cart
22-651-181 View Documents Nor-Lake™ Scientific
PHPF021WWW/0
Automatic 1.3 cu. ft. Solid Swing Each for $1,661.52
Add to cart
22-651-182 View Documents Nor-Lake™ Scientific
PHPF021WWWLH/0
Automatic 1.3 cu. ft. Solid swing door-Left Hinged Each for $1,748.21
Add to cart
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • 2 probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts
  • Freestanding thermometer with 3 year certificate of calibration
  • Keyed door locks, leveling legs, and pharmacy refrigerator/freezer toolkit
Specifications
Specifications
Enameled Steel
1
NEMA 5-15 P
60 Hz
R600a
Digital
1 ph
High density urethane foam
Audible and visual high and low temperature alarms
115 V