Learn More
Nor-Lake™ Scientific Premier Pharmacy Freestanding Undercounter Refrigerator
Ideal choice for pharmacies with undercounter and counter top applications with adequate space for ventilation.
$986.48 - $1996.00
Specifications
Defrost Cycle
Display Digital
Ports 1
Phase 1 ph
Plug Type NEMA 5-15 P
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
22-651-172
View Documents
Nor-Lake™ Scientific
PHPR011WWW/0
1 cu. ft. Single Solid Swing
Each for $1,829.00
Add to cart
22-651-173
View Documents
Nor-Lake™ Scientific
PHPR011WWWLH/0
1 cu. ft. Single Solid Swing Left-Hinged
Each for $986.48
Add to cart
22-651-174
View Documents
Nor-Lake™ Scientific
PHPR031WWW/0
2.5 cu. ft. Single Solid Swing
Each for $1,893.00
Add to cart
22-651-175
View Documents
Nor-Lake™ Scientific
PHPR011WWG/0
1 cu. ft. 1 Glass Swing Door
Each for $1,850.00
Add to cart
22-651-176
View Documents
Nor-Lake™ Scientific
PHPR011WWGLH/0
1 cu. ft. Single Glass Swing Left-Hinged
Each for $1,863.00
Add to cart
22-651-177
View Documents
Nor-Lake™ Scientific
PHPR031WWG/0
2.5 cu. ft. 1 Glass Swing Door
Each for $1,893.00
Add to cart
22-651-178
View Documents
Nor-Lake™ Scientific
PHPR061WWG/0
5.2 cu. ft. 1 Glass Swing Door
Each for $1,996.00
Add to cart
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • 2 probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts
  • Freestanding thermometer with 3 year certificate of calibration
  • Keyed door locks, leveling legs, and pharmacy refrigerator/freezer toolkit temperature logs and CDC approved power cord labels
Specifications
Specifications
Cycle
1
NEMA 5-15 P
60 Hz
R600a
115 V
Digital
1 ph
High density urethane foam
Audible and visual high and low temperature alarms
Microprocessor