Learn More
Nor-Lake™ Scientific Premier Pharmacy Stainless Steel Laboratory Series Refrigerator
Nor-Lake™ Scientific Premier Pharmacy Stainless Steel Laboratory Series Refrigerator feature microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats.
$4271.00 - $7042.00
Specifications
Cabinet Material Stainless Steel
Casters Swivel casters
Defrost Cycle
Display Digital
Door Style Stainless Steel Swing
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Refrigerator Temperature Range (Refrigerator) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Refrigerator Temperature Range (Refrigerator) Price Quantity & Availability  
22-651-154
View Documents
Nor-Lake™ Scientific
PHPR231SSS/0
23 cu. ft.
Each for $4,271.00
Add to cart
22-651-155
View Documents
Nor-Lake™ Scientific
PHPR492SSS/0
49 cu. ft.
Each for $7,042.00
Add to cart
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • Freestanding thermometer with 3 years certificate of calibration
  • Two probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts, keyed door locks, casters, and pharmacy refrigerator toolkit, temperature logs and CDC approved power cord labels
Specifications
Specifications
Stainless Steel
Cycle
Stainless Steel Swing
60 Hz
Audible and visual high and low temperature alarms
1 ph
1
Microprocessor
Swivel casters
Digital
Stainless Steel
High density urethane foam
115 V
NEMA 5-15 P
R134a