Learn More
Nor-Lake™ Scientific Premier Solid Door Series Pharmacy Refrigerator, Bottom Mount
Nor-Lake™ Scientific Premier Solid Door Series Pharmacy Refrigerator feature all-new temperature display and alarm module offering array of robust security features with monitoring and alarm system, independent of microprocessor temperature controller for additional product security.
$5761.00 - $13800.00
Specifications
Casters Yes
Defrost Cycle
Display Digital
Frequency 60 Hz
Insulation Foam insulation
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
22-651-208
View Documents
Nor-Lake™ Scientific
PHPR121WWW/0
12 cu. ft. Solid Swing
Each for $5,761.00
Add to cart
22-651-209
View Documents
Nor-Lake™ Scientific
PHPR161WWW/0
16 cu. ft. Solid Swing
Each for $6,791.00
Add to cart
22-651-210
View Documents
Nor-Lake™ Scientific
PHPR231WWW/0
23 cu. ft. Solid Swing
Each for $7,345.00
Add to cart
22-651-211
View Documents
Nor-Lake™ Scientific
PHPR261WWW/0
26 cu. ft. Solid Swing
Each for $7,547.00
Add to cart
22-651-212
View Documents
Nor-Lake™ Scientific
PHPR362WWW/0
36 cu. ft. Solid Swing
Each for $9,410.00
Add to cart
22-651-213
View Documents
Nor-Lake™ Scientific
PHPR492WWW/0
49 cu. ft. Solid Swing
Each for $10,808.00
Add to cart
22-651-214
View Documents
Nor-Lake™ Scientific
PHPR723WWW/0
72 cu. ft. Solid Swing
Each for $13,800.00
Add to cart
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • Two probes, 1 in air and 1 in sample bottle
  • Remote alarm contacts
  • Freestanding thermometer with 3 years certificate of calibration
  • Keyed door locks, casters, and pharmacy refrigerator toolkit, temperature logs and CDC approved power cord labels
Specifications
Specifications
Yes
Digital
Foam insulation
1 ph
1
Microprocessor
Cycle
60 Hz
Audible and visual high and low temperature alarms
NEMA 5-15 P
R134a
115 V