Learn More
ML Kishigo Premium Black Series™ Surveyors Vest
ML Kishigo Premium Black Series™ Surveyors Vest with ANSI 107 Class 2 Safety Compliance
Manufacturer:  ML Kishigo S50025XL
 View more versions of this product
Catalog No. 19-441-119
Add to cart
Description
Description
  • Solid polyester front with mesh back
  • Heavy duty black bottom to help maintain a clean appearance
  • Zipper front closure reinforced with durable webbing
  • 2 in. wide high performance reflective material laminated to 3 in. contrasting color
  • Left and right mic tabs (Size small only has left mic tab)
  • 2 lapel grommets
  • Padded neck for comfort
Specifications
Specifications
ANSI 107 Class 2
Lime
Polyester
79 cm
Solid Front, Mesh Back
5X-Large