Learn More
Eisco™ Premium Die Casted Boss Head Clamp
Ideal for connecting 16mm diameter rods and apparatus in tight places without sacrificing strength.
Manufacturer:  Eisco™ PRBH01/DEL
Catalog No. 04-999-146
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Threaded metal one piece clamping screws
  • All metal constructions to carry heavy loads and prevent torque damage
  • Able to hold rods up to 16mm in diameter
  • Holds rods orthogonal (90 degrees) to one another
  • Made from high grade zinc alloy with non toxic powder coating finish
  • With two stainless steel thumb screws