Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Premium Dynamometers (Spring Scales)
SCI_ED

Premium Dynamometers feature internal red cylinders for easy and accurate measurments.

$11.50 - $18.00

Specifications

Height (Metric) 25 cm
Includes 2 hooks, Spring in acrylic tube
Weight (Metric) 0.09 oz
Width (Metric) 2.5 cm
View More Specs

Products 10
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
S88331
SCI_ED
View Documents
Eisco™
PH0036PRA
10 g, 0.1 N
Each for $13.00
Add to Cart
 
S88332
SCI_ED
View Documents
Eisco™
PH0036PRB
20 g, 0.2 N
Each for $11.50
Add to Cart
 
S88333
SCI_ED
View Documents
Eisco™
PH0036PRC
100 g, 1 N
Each for $16.25
Add to Cart
 
S88334
SCI_ED
View Documents
Eisco™
PH0036PRD
200 g, 2 N
Each for $13.00
Add to Cart
 
S88335
SCI_ED
View Documents
Eisco™
PH0036PRD01
250 g, 2.5 N
Each for $13.00
Add to Cart
 
S88336
SCI_ED
View Documents
Eisco™
PH0036PRE
500 g, 5 N
Each for $13.00
Add to Cart
 
S88337
SCI_ED
View Documents
Eisco™
PH0036PRF
1 kg, 10 N
Each for $18.00
Add to Cart
 
S88338
SCI_ED
View Documents
Eisco™
PH0036PRG
2 kg, 20 N
Each for $15.50
Add to Cart
 
S88339
SCI_ED
View Documents
Eisco™
PH0036PRH
5 kg, 50 N
Each for $13.00
Add to Cart
 
S88340
SCI_ED
View Documents
Eisco™
PH0036PRI
10 kg, 100 N
Each for $15.00
Add to Cart
 
Description

Description

  • Accurate and precise springs with excellent linearity
  • All metal parts encased in a premium acrylic tube
  • Enclosed springs prevent tangling or loss of functionality
  • Twist knob to zero out
  • Features a thick clear acrylic wall
  • Color coded according to measurement range

Warranty and Services

1 year limited manufacturer warranty

Specifications

Specifications

25 cm
2 hooks, Spring in acrylic tube
0.09 oz
2.5 cm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit