Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Unibor™ Premium Gel Hand Sanitizer

Gel hand sanitizer with green tea extract is effective, safe, and leaves your hands feeling soft. Trusted by hospitals, schools, and governments worldwide.

Specifications

Certifications/Compliance FDA Registered OTC Monograph, Part333A
Color Clear
For Use With (Application) Eliminating Germs on Hands
Fragrance Neutral
Product Type Gel Hand Sanitizer
View More Specs

Alert:

  • Not for consumption
  • Flammable


Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-350-102
View Documents
Unibor™
UHS338
3.4 oz. Case of 60 for $240.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-350-103
View Documents
Unibor™
UHS236
8 oz. Case of 10 for $60.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-350-104
View Documents
Unibor™
UHS500
17 oz. Case of 30 for $228.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-350-105
View Documents
Unibor™
UHS1G
1 Gallon Case of 4 for $170.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Regulary tested for effectiveness against pathogens
  • Produced with green tea extract and other moisturizing ingredients
  • Fully registered with the US FDA, Korean KFDA, EU, Health Canada, and many more
  • Free of harmful ingredients - tested annually by independent labs
  • Available in small flip cap bottles and 3 pump bottle sizes
Specifications

Specifications

FDA Registered OTC Monograph, Part333A
Eliminating Germs on Hands
Gel Hand Sanitizer
Clear
Neutral
Gel
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit