Learn More
Eisco™ Premium Large Three Finger Clamp
Choose durable lab equipment that will last for years. The EISCO Premium Large Three Finger Clamp is an industrial clamp that has rubber coated fingers.
Manufacturer:  Eisco™ PRCMP07/DEL
Catalog No. 04-999-140
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Clamp constructed of powder coated steel, 3 finger rubber coated clamp attached to aluminum rod
  • Maximum clamp opening is 8.5cm
  • 14cm connecting rod
  • Completely metal constructions
  • Thread fitted actuating joints
  • 10mm diameter connecting rod