Learn More
Gilson™ Premium Pipette Service for Multichannel Pipettes
Gilson™ Premium Pipette Service is the most comprehensive level of service, providing users with reliable performance and predictable pricing. At least once per year Gilson comes with a lifetime warranty on the pipette and all necessary repairs are included at no additional cost.
Supplier: Gilson™ SVC101228
Description
- Visual and Functional Inspection
- Internal and External Cleaning
- Leak Test
- Gravimetric Check
- Preventative Maintenance
- Re-grease as needed
- Pipette Label
- Mail-In or Onsite Service Available
- Calibration and Detailed Certificate
- ISO 17025 Calibration Available
- Lifetime Warranty
- Free Repairs
- 3 day turn around time
Specifications
12-channel Pipettes | |
ISO 17025 Accredited Service | |
3 Days |
4 data points collected from all channels at 2 volumes | |
Data collected before and after repair | |
Pipette Premium Lifetime Service |