Learn More
Sartorius™ Premium Pro20 Benchtop Conductivity Meters
Exact measurements with wider measuring ranges and better accuracy specifications
Manufacturer:  Sartorius™ PR20CONKIT/DEL
 View more versions of this product
Catalog No. 14-557-885
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Pro20
0 to 1999mg/L
0.0 to 70.0 (acc. to IOT); 0.00 to 20 MOhm cm
0.0° to 100°C
±0.1°C
500 data sets
50/60Hz
External Universal Power Supply: primary: 100 to 240V, 50/60Hz; secondary: 9V = 1.5A
0.000μS/cm to 1000mS/cm
0.01
Automatic
0.1
LCD
CE marked
100 to 240V
19cm