Learn More
Eisco™ Premium Square Head Boss Head Clamp
Able to hold rods up to 15mm in diameter.
Manufacturer:  Eisco™ PRBH05/DEL
Catalog No. 04-999-150
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Threaded metal one piece clamping screws
  • Able to hold rods up to 15mm in diameter
  • Holds rods orthogonal (90 degrees) to one another
  • All metal constructions to carry heavy loads and prevent torque damage/li>