Learn More
Eisco™ Premium Three Finger Clamp
Choose durable lab equipment that will last for years. The EISCO Premium Three Finger Clamp is an industrial clamp with rubber coated fingers.
Manufacturer:  Eisco™ PRCMP01/DEL
Catalog No. 04-999-132
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Clamp constructed of powder coated steel
  • Maximum clamp opening is 8.5cm
  • 14cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10mm diameter connecting rod