Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Premium Two Prong Clamp w/ Boss Head Attachment

Choose durable lab equipment that will last for years. The EISCO Premium Two Prong Clamp w/ Boss Head Attachment is an industrial,rubber-coated clamp for burettes, flasks, and beaker clamps with a swivel boss head attachment.

Manufacturer:  Eisco™ PRCMP05BH/DEL

Catalog No. 04-999-137

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »

This item is not returnable. View return policy

Description

Description

  • Maximum clamp opening is 10.5 cm
  • Swivel between clamp and boss head
  • Boss accommodates to 19mm rod
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit