Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Premium Two Prong Clamp w/ Boss Head Attachment

Choose durable lab equipment that will last for years. The EISCO Premium Two Prong Clamp w/ Boss Head Attachment is an industrial, cork-lined clamp for burettes, flasks, and beakers. It features a swivel head boss head attachment.

Supplier:  Eisco™ PRCMP08BH/DEL

Catalog No. 04-999-143

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • Maximum clamp opening is 10.5 cm
  • Swivel between clamp and boss head
  • Boss accommodates to 19mm rod
Specifications

Specifications

Not Autoclavable
Blue
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit