Learn More
Eisco™ Premium Two Prong Clamp w/ Dual Screw Adjustments
Choose durable lab equipment that will last for years. The EISCO Premium Two Prong Clamp w/ Dual Screw Adjustments is an industrial, rubber-coated lab clamp with dual screw adjustments.
Manufacturer:  Eisco™ PRCMP04/DEL
Catalog No. 04-999-159
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Heavy duty spring loaded clamp jaw with dual screw adjustment to fine tune any fitting
  • Made of steel and powder coated alloy with all metal adjustment screws
  • 2 rubber coated clamp tongs
  • 7cm maximum clamp opening
  • 12.5cm connection rod