Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Premixed Digestion Fluid, 50% HCl / 2% HNO3, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner SP07520020F

Catalog No. SP07520020F


Due to product restrictions, please to purchase or view availability for this product.


Specifications

Specifications

Digestion Fluid
7647-01-0, 7732-18-5, 7697-37-2
54.0, 45.53, 2.74
Liquid
Colorless to Green-Yellow
Premixed
52.0, 43.75, 2.63
50% HCl / 2% HNO3
Cubitainer
20L
Safety and Handling

Safety and Handling

ShelfLife : 24 months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit