Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PREP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556724
Description
Immunogen sequence: LVKYSPLHNV KLPEADDIQY PSMLLLTADH DDRVVPLHSL KFIATLQYIV GRSRKQSNPL LIHVDTKAGH GAGKPTAKVI EEVSDMFAFI ARCLNVDWIP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
Specifications
PREP | |
Polyclonal | |
Unconjugated | |
PREP | |
AI047692; AI450383; D10Wsu136e; dJ355L5.1 (prolyl endopeptidase); EC 3.4.21.26; MGC16060; PE; PE {ECO:0000250; PEP; Pop; post-proline cleaving enzyme; post-proline cleaving enzyme {ECO:0000250; Prep; prep {ECO:0000312; Prolyl endopeptidase; prolyl endopeptidase {ECO:0000250; prolyl oligopeptidase; RGD:620841}; rPop; UniProtKB:P23687} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5550 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48147 | |
PREP | |
Recombinant protein corresponding to Human PREP. Recombinant protein control fragment (Product #RP-94644). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction