Learn More
SSIbio Preparation Racks for PCR Thin-Walled Tubes
For PCR preparation and storage
Manufacturer:  SSIbio 523021
 View more versions of this product
Catalog No. 05-541-50
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Racks with covers designed for preparing and storing 0.2mL thin-walled tubes and strip caps before or after PCR sample processing applications
  • Racks include alphanumeric index for easy tube identification
  • Transparent plastic covers have a recessed lid that allows racks to be stacked
  • Compatible with other manufacturers' tray/retainer sets or cycler plates, individual tubes and 8- or 12-strip tubes
Capacity: 96 positions (8 x 12 format) for 0.2mL thin-walled tubes and strip caps.
Specifications
Specifications
Not Autoclavable
Blue
Microtube Rack