Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prepared Culture Media in Plates

Labeled with product name, expiration date and lot number.
Supplier: Fischer Technical Company CM115P25
Description
- Sterile plates are ready for immediate use
- Supplied in disposable plastic Petri dishes, 100mm O.D. × 15mm deep
- Packed in clear, heat-sealed cellophane bags
Specifications
2°C to 8°C | |
US | |
Sterile | |
Petri Dish | |
0.59 in. | |
3.93 in. |
Plate | |
Brain heart infusion Agar culture media | |
Cultivation of wide variety of fastidious and non-fastidious microorganisms | |
10/Pk. | |
15 mm | |
100 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title