Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prepared Culture Media in Tubes
Ready-to-use media prepared in disposable borosilicate glass tubes.
$27.50 - $94.00
Specifications
Packaging | Glass tube with black screw cap |
---|---|
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Media Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Media Type | Quantity | Price | Quantity & Availability | ||||
S716932A
![]() |
Fischer Technical Company
CM145T1020 |
Nutrient Agar, Tube | 20 Tubes |
Pack of 20 for $89.00
|
|
S716942A
![]() |
Fischer Technical Company
CM146T1020 |
Nutrient Broth | 20 Tubes |
Pack of 20 for $94.00
|
|
S716933A
![]() |
Fischer Technical Company
CM145S920 |
Nutrient Agar, Slant | 20 Tubes |
Pack of 20 for $59.75
|
|
S716992A
![]() |
Fischer Technical Company
CM164T1020 |
Tryptic Soy Broth | 20 Tubes |
Pack of 20 for $83.00
|
|
S716972A
![]() |
Fischer Technical Company
CM150T1020 |
Sabouraud Dextrose Agar | 20 Tubes |
Pack of 20 for $27.50
|
|
Description
- Tubes measure 16mm O.D. x 125mm long and have black plastic screw caps
- Each tube is labeled with product name, expiration date and lot number
- Refrigerate until ready to use
Order Info
Sold in pack of 20 tubes.
Specifications
Glass tube with black screw cap | |
United States |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title