Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Prepared Culture Media in Tubes
SCI_ED

Ready-to-use media prepared in disposable borosilicate glass tubes.

Supplier:  Fischer Technical Company™ CM145T1020

 View more versions of this product

Catalog No. S716932A


Add to Cart

Alert:

Prop 65: Cancer and Reproductive Harm.


Description

Description

  • Tubes measure 16 mm O.D. x 125 mm long
  • Include black plastic screw caps
  • Each tube is labeled with product name, expiration date and lot number
  • Refrigerate until ready to use

Order Info

Sold in pack of 20 tubes.

Specifications

Specifications

US with Globally Sourced Materials
Nutrient Agar
10 mL
20 Tubes/Pk.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit