Learn More
Presenilin-1 Mouse, DyLight 488, Clone: 34-4C3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178992G
Catalog No. NBP178992G
Add to cart
Description
Description
Presenilin-1 Monoclonal antibody specifically detects Antigen in Human, Mouse, Primate, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blotting.
Specifications
Specifications
Presenilin-1
34-4C3
Western Blot, Immunohistochemistry-Frozen
Purified
PSEN1
The I3 loop of the M2 receptor fusion protein (residues 225-359) was used as an immunogen
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Primate, Rat
Immunohistochemistry (Frozen), Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182
Mouse
IgG1
0.1 ml
Alzheimers Research, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers, Stem Cell Signaling Pathway, Tumor Suppressors, Wnt Signaling Pathway
Primary
5663