Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Presenilin-1 Rat, DyLight 350, Clone: 24-4B5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP178993UV

Catalog No. NBP178993UV


Add to cart

Description

Description

Presenilin-1 Monoclonal antibody specifically detects Antigen in Human, Mouse, Rat samples. It is validated for Western Blotting.
Specifications

Specifications

Presenilin-1
24-4B5
Western Blot, Immunohistochemistry
Purified
PSEN1
Recombinant peptide corresponding to the N-terminus of Presenilin 1
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182
Rat
IgG1
0.1 ml
Alzheimers Research, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers, Stem Cell Signaling Pathway, Tumor Suppressors, Wnt Signaling Pathway
Primary
5663
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit