Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Pressure Filtration Connections
Inlet/Outlet Connections for MD 050 and MD 142/5 Stainless Steel Pressure Filtration Devices
$14.44 - $77.43
Specifications
Material Stainless Steel
Includes PTFE Seal
For Use With (Equipment) Series MD 050, MD 142/5 Pressure Filtration Devices
View More Specs
Products 2
Catalog Number Mfr. No. Connector Types Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Price Quantity & Availability  
09-920-142
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10453007
Olive External Diameter 9 to 11 mm Each for $14.44
Add to cart
09-920-139
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10453001
Rapid Seal Coupling Each for $77.43
Add to cart
Specifications
Specifications
Stainless Steel
PTFE Seal
Series MD 050, MD 142/5 Pressure Filtration Devices