Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ PrestoBlue™ Cell Viability Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.

PrestoBlue™ Cell Viability Reagent

$269.00 - $552.00

Specifications

Color Blue
Concentration 10 x
Detection Method Colorimetric, fluorescent
Excitation Wavelength Range 535 to 560nm
For Use With (Application) For cell counting, viability, and cryopreservation, cell viability, proliferation and function
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A13261 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Molecular Probes™
A13261
25mL Each for $269.00
Add to cart
 
A13262 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Molecular Probes™
A13262
100mL Each for $552.00
Add to cart
 
Description

Description

Get high quality cell viability results and instant time savings with PrestoBlue™ Cell Viability Reagent. PrestoBlue™ is a ready to use cell permeable resazurin-based solution that functions as a cell viability indicator by using the reducing power of living cells to quantitatively measure the proliferation of cells. When added to cells, the PrestoBlue™ reagent is modified by the reducing environment of the viable cell and turns red in color, becoming highly fluorescent. This color change can be detected using fluorescence or absorbance measurements.

The PrestoBlue™ Cell Viability Reagent allows you to:

  • Save time with an incubation step as short as 10 minutes.
  • Obtain high quality results with a large dynamic range
  • Experience convenience with the add and read homogenous format
  • Monitor living cells and perform downstream functional assays

    10-minute Incubations
    The PrestoBlue™ Cell Viability Reagent is the only live-cell viability reagent on the market able to provide robust data typically in as little as 10 minutes following reagent addition. All other resazurin-based cell viability reagents on the market require a one to four hour incubation with the reagent prior to obtaining data.

    High Quality Results
    Based on the cell type and incubation time, PrestoBlue™ Cell Viability Reagent can detect as few as 12 cells per well in a 384 well plate. After ten minutes of incubation the PrestoBlue™ Reagent can linearly detect cells down to 98 cells⁄well with a Z’ value of greater than 0.5.

    Homogenous Assay Format
    The PrestoBlue™ Cell Viability Reagent is an addition-only reagent. Without the cell lysis or solubilization steps required by other assays on the market, hands-on time is minimized.

    Live Cell Assay
    The PrestoBlue™ Cell Viability Reagent does not require cell lysis. Cells may be incubated for up to 24 hours in the presence of PrestoBlue™ Cell Viability Product. Simply remove the reagent from the cells and replace it with growth medium for further proliferation.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Specifications

    Specifications

    Blue
    Colorimetric, fluorescent
    For cell counting, viability, and cryopreservation, cell viability, proliferation and function
    384-well plate, 96-well plate, Cuvettes, Liquid
    PrestoBlue™ Cell Viability Reagent
    10 x
    535 to 560nm
    For HTS reader, microplate reader, spectrophotometer
    2 to 8°C

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit