Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Supplier:  Thermo Scientific™ 50134022
 View more versions of this product
Catalog No. 10-451-222
Add to Cart
Description
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
Specifications
5µm filter with carbon with 10 ft. filter housing
Filter
3.048 m
LabTower EDI, LabTower RO, LabTower TII
10 ft.